Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040667.2 | NP_001035757.1 | 10 | 1712 | |
NM_001538.3 | NP_001529.2 | 10 | 1726 | ||
GenBank mRNA | AB029348.1 | - | 8 | 754 | |
AK296210.1 | - | 8 | 769 | ||
BC130383.1 | - | 8 | 815 | ||
D87673.1 | - | 8 | 768 |