Gene Transcripts

Gene Symbol
Msi1
Entrez Gene ID
17690
Gene Name
musashi RNA-binding protein 1
Gene Aliases
Msi1h, Musahi1, m-Msi-1
Chromosome Location
Chr. 5: 115359083 - 115455698 on Build GRCm38
UniGene ID
Mm.391995
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008629.1 NP_032655.1 7, 8 523
GenBank mRNA AK084019.1 - 7, 8 506
D49654.1 - 7, 8 523

View Full Product Details