Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008751.5 | NP_032777.3 | 2 | 356 | |
GenBank mRNA | AK043942.1 | - | 2 | 561 | |
AK047847.1 | - | 3 | 591 | ||
AK083014.1 | - | 3 | 935 | ||
AK163438.1 | - | 3 | 383 | ||
BC068137.1 | - | 2 | 699 | ||
BC079608.1 | - | 2 | 649 | ||
BC145953.1 | - | 2 | 356 | 40131256 | |
BC145955.1 | - | 2 | 356 | 40131258 |