Gene Transcripts

Gene Symbol
P2rx1
Entrez Gene ID
18436
Gene Name
purinergic receptor P2X, ligand-gated ion channel, 1
Gene Aliases
AI323649, BB122383, P2x, Pdcd3, RP-2
Chromosome Location
Chr. 11: 72999056 - 73015200 on Build GRCm38
UniGene ID
Mm.25722
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008771.3 NP_032797.3 4, 5 598
GenBank mRNA AK035304.1 - 4, 5 599
BC015084.1 - 4, 5 612 4189541
X84896.1 - 4, 5 592

View Full Product Details