Gene Transcripts

Gene Symbol
Pbx3
Entrez Gene ID
18516
Gene Name
pre B cell leukemia homeobox 3
Gene Aliases
Chromosome Location
Chr. 2: 34171457 - 34372045 on Build GRCm38
UniGene ID
Mm.239941
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290576.1 NP_001277505.1 7 1151
NM_016768.2 NP_058048.1 8 1264
GenBank mRNA AF020199.1 - 8 1255
AF020200.1 - 7 1144
AK086310.1 - 1 3377
AK144363.1 - 7 1068
BC057879.1 - 8 1155 5348692

View Full Product Details