Gene Transcripts
- Gene Symbol
- Pkm
- Entrez Gene ID
- 18746
- Gene Name
- pyruvate kinase, muscle
- Gene Aliases
- AA414905, AL024370, AL024424, Pk-2, Pk-3, Pk3, Pkm2
- Chromosome Location
- Chr. 9: 59656368 - 59679375 on Build GRCm38
- UniGene ID
- Mm.326167
- Species
- Mus musculus
- Gene Symbol
- C330027C09Rik
- Entrez Gene ID
- 224171
- Gene Name
- RIKEN cDNA C330027C09 gene
- Gene Aliases
- AA408511, AU018569, Cip2a, Kiaa1524
- Chromosome Location
- Chr. 16: 48994188 - 49019711 on Build GRCm38
- UniGene ID
- Mm.24491
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC021874.1 |
- |
|
16 |
|
View Full Product Details