Gene Transcripts

Gene Symbol
Ppox
Entrez Gene ID
19044
Gene Name
protoporphyrinogen oxidase
Gene Aliases
Ppo
Chromosome Location
Chr. 1: 171276991 - 171281203 on Build GRCm38
UniGene ID
Mm.266888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008911.2 NP_032937.1 7 1053
GenBank mRNA AK134341.1 - 1557
BC002047.1 - 7 1008 3484079
D45185.1 - 7 779
U25114.1 - 7 1028

View Full Product Details