Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
19763
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr. 17: 34020792 - 34024680 on Build GRCm38
UniGene ID
Mm.20343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009066.3 NP_033092.3 5, 6 1072
GenBank mRNA AK154962.1 - 5, 6 1033
AK155509.1 - 5, 6 1048
BC009070.1 - 5, 6 1035 2651194
BU918502.1 - 1, 2 115
CT010219.1 - 3, 4 762
KJ650216.1 - 4, 5 849
Y12881.1 - 3, 4 768

View Full Product Details