Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000640.2 | NP_000631.1 | 8, 9 | 1130 | |
GenBank mRNA | AK291957.1 | - | 8, 9 | 1099 | |
AK308523.1 | - | 8, 9 | 1061 | ||
AK309444.1 | - | 8, 9 | 1140 | ||
BC020739.1 | - | 7, 8 | 1031 | 4807603 | |
BC033705.2 | - | 7, 8 | 1039 | 3876064 | |
U70981.1 | - | 8, 9 | 1098 | ||
X95302.1 | - | 8, 9 | 1057 | ||
Y08768.1 | - | 8, 9 | 1130 |