Gene Transcripts
- Gene Symbol
- Stfa1
- Entrez Gene ID
- 20861
- Gene Name
- stefin A1
- Gene Aliases
- Stefin-3, Stf-3, Stf1, Stf3, Stfa3
- Chromosome Location
- Chr. 16: 36277148 - 36285371 on Build GRCm38
- UniGene ID
- Mm.327618
- Species
- Mus musculus
- Gene Symbol
- BC100530
- Entrez Gene ID
- 100034684
- Gene Name
- cDNA sequence BC100530
- Gene Aliases
- Chromosome Location
- Chr. 16: 36359382 - 36367570 on Build GRCm38
- UniGene ID
- Mm.383561
- Species
- Mus musculus
- Gene Symbol
- BC117090
- Entrez Gene ID
- 100038854
- Gene Name
- cDNA sequence BC1179090
- Gene Aliases
- Gm1975
- Chromosome Location
- Chr. 16: 36321665 - 36334332 on Build GRCm38
- UniGene ID
- Mm.383561
- Species
- Mus musculus
View Full Product Details