Gene Transcripts

Gene Symbol
Stfa1
Entrez Gene ID
20861
Gene Name
stefin A1
Gene Aliases
Stefin-3, Stf-3, Stf1, Stf3, Stfa3
Chromosome Location
Chr. 16: 36277148 - 36285371 on Build GRCm38
UniGene ID
Mm.327618
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082543.1 NP_001076012.1 2, 3 227
GenBank mRNA BY113926.1 - 2, 3 227
M92417.1 - 2, 3 196

Gene Symbol
BC100530
Entrez Gene ID
100034684
Gene Name
cDNA sequence BC100530
Gene Aliases
Chromosome Location
Chr. 16: 36359382 - 36367570 on Build GRCm38
UniGene ID
Mm.383561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082546.1 NP_001076015.1 2, 3 248
GenBank mRNA BC100530.1 - 2, 3 426

Gene Symbol
BC117090
Entrez Gene ID
100038854
Gene Name
cDNA sequence BC1179090
Gene Aliases
Gm1975
Chromosome Location
Chr. 16: 36321665 - 36334332 on Build GRCm38
UniGene ID
Mm.383561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001332.2 NP_001001332.2 2, 3 215
GenBank mRNA AY169235.1 - 2, 3 215
BC117064.1 - 2, 3 215 40126383
BC117090.1 - 2, 3 215 40126409

View Full Product Details