Gene Transcripts

Gene Symbol
Stk11
Entrez Gene ID
20869
Gene Name
serine/threonine kinase 11
Gene Aliases
AA408040, Lkb1, Par-4, R75140
Chromosome Location
Chr. 10: 80115693 - 80130685 on Build GRCm38
UniGene ID
Mm.44231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB015801.1 - 8 1015
AF129870.1 - 8 1393
AF145287.1 - 8 997
AF151711.1 - 8 997
AK004499.1 - 6 676
AK147802.1 - 8 1057
AK171909.1 - 9 1791
AK172385.1 - 8 1780
AK172528.1 - 8 1777
BC052379.1 - 8 1058 5688695
EU730638.1 - 8 976

View Full Product Details