Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011495.2 | NP_035625.2 | 6 | 1565 | |
NM_173169.2 | NP_775261.2 | 6 | 1565 | ||
GenBank mRNA | AK006459.1 | - | 2 | 94 | |
AK045082.1 | - | 6 | 1489 | ||
AK137080.1 | - | 6 | 1477 | ||
AK137471.1 | - | 6 | 1476 | ||
AK164465.1 | - | 6 | 1471 | ||
BC026785.1 | - | 6 | 1475 | 5150173 | |
BC057940.1 | - | 6 | 1462 | 5323295 | |
L29479.1 | - | 6 | 1467 |