Gene Transcripts
- Gene Symbol
- INSRR
- Entrez Gene ID
- 3645
- Gene Name
- insulin receptor related receptor
- Gene Aliases
- IRR
- Chromosome Location
- Chr. 1: 156840873 - 156858920 on Build GRCh38
- UniGene ID
- Hs.248138
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014215.2 |
NP_055030.1
|
20 |
3711 |
|
GenBank mRNA |
J05046.1 |
- |
19 |
3327 |
|
View Full Product Details