Gene Transcripts
- Gene Symbol
- Vasp
- Entrez Gene ID
- 22323
- Gene Name
- vasodilator-stimulated phosphoprotein
- Gene Aliases
- Chromosome Location
- Chr. 7: 19256929 - 19271854 on Build GRCm38
- UniGene ID
- Mm.9684
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282021.1 |
NP_001268950.1
|
2, 3 |
518 |
|
NM_001282022.1 |
NP_001268951.1
|
2, 3 |
518 |
|
NM_009499.3 |
NP_033525.2
|
2, 3 |
518 |
|
GenBank mRNA |
AF084548.1 |
- |
2, 3 |
485 |
|
AK140329.1 |
- |
2, 3 |
482 |
|
AK150103.1 |
- |
2, 3 |
480 |
|
AK155480.1 |
- |
2, 3 |
480 |
|
AK157021.1 |
- |
2, 3 |
482 |
|
AK170780.1 |
- |
2, 3 |
474 |
|
AK171302.1 |
- |
2, 3 |
482 |
|
AK171715.1 |
- |
2, 3 |
482 |
|
BC010223.1 |
- |
2, 3 |
220 |
3500587 |
BC015289.1 |
- |
2, 3 |
461 |
4240907 |
View Full Product Details