Gene Transcripts

Gene Symbol
Ly6h
Entrez Gene ID
23934
Gene Name
lymphocyte antigen 6 complex, locus H
Gene Aliases
Chromosome Location
Chr. 15: 75564745 - 75567249 on Build GRCm38
UniGene ID
Mm.22154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135688.1 NP_001129160.1 4 400
NM_001135689.1 NP_001129161.1 4 314
NM_011837.3 NP_035967.1 4 677
GenBank mRNA AF127091.1 - 4 310
AK021002.1 - 4 354
AK034884.1 - 317
BB627076.1 - 4 317
BC028758.1 - 4 311 1245940
BM943584.1 - 4 400
BM948855.1 - 4 677
CX238172.1 - 4 380

View Full Product Details