Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164406.1 | NP_001157878.1 | 5 | 434 | |
NM_025291.3 | NP_079567.2 | 5 | 537 | ||
GenBank mRNA | AK007302.1 | - | 5 | 523 | |
AK010375.1 | - | 5 | 523 | ||
AK154232.1 | - | 5 | 507 | ||
BC026480.1 | - | 3 | 316 | ||
BC048362.1 | - | 5 | 509 | 2646489 | |
BQ748094.1 | - | 3 | 205 |