Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008580.4 | NP_032606.4 | 2 | 610 | |
GenBank mRNA | AB006787.3 | - | 2 | 662 | |
BC116627.1 | - | 2 | 610 | 40086293 | |
BC116628.1 | - | 2 | 610 | 40086299 | |
BC133697.1 | - | 2 | 610 | 40086283 |