Gene Transcripts

Gene Symbol
Cops7a
Entrez Gene ID
26894
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 7a (Arabidopsis thaliana)
Gene Aliases
D6Ertd35e
Chromosome Location
Chr. 6: 124958411 - 124965529 on Build GRCm38
UniGene ID
Mm.244536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164089.1 NP_001157561.1 2 320
NM_012003.2 NP_036133.1 2 305
GenBank mRNA AF071316.1 - 2 307
AF240179.1 - 1 125
AK008474.1 - 2 311
AK010417.1 - 2 315
AK010726.1 - 2 290
AK013155.1 - 2 273
AK019122.1 - 2 297
BC003724.1 - 1 186 3593979
BG085353.2 - 2 305
CJ165979.1 - 2 301

View Full Product Details