Gene Transcripts

Gene Symbol
Nit1
Entrez Gene ID
27045
Gene Name
nitrilase 1
Gene Aliases
AI255805, ESTM30, W57327
Chromosome Location
Chr. 1: 171342237 - 171345645 on Build GRCm38
UniGene ID
Mm.12915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242580.1 NP_001229509.1 3 390
NM_012049.2 NP_036179.1 3 358
GenBank mRNA AF069988.1 - 3 339
AK002269.1 - 3 397
AK004988.2 - 351
AK032189.1 - 921
AK200650.1 - 2 238
AK205756.1 - 2 238
BC021634.1 - 3 340 4008543

View Full Product Details