Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016772.1 | NP_058052.1 | 3 | 312 | |
GenBank mRNA | AF030343.1 | - | 3 | 312 | |
AK088495.1 | - | 3 | 300 | ||
BC068112.1 | - | 3 | 312 | 5710971 | |
BC087924.1 | - | 3 | 373 | 6706361 |