Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290824.1 | NP_001277753.1 | 11 | 1382 | |
NM_153387.3 | NP_700436.1 | 11 | 1382 | ||
GenBank mRNA | AK034694.1 | - | 2 | 912 | |
AK038921.1 | - | 11 | 1341 | ||
AK045305.1 | - | 11 | 1275 | ||
AK046132.1 | - | 11 | 1312 | ||
AK050250.1 | - | 11 | 1383 | ||
AK051517.1 | - | 11 | 1283 | ||
AK152984.1 | - | 11 | 1341 | ||
AK153171.1 | - | 11 | 1341 | ||
AK161774.1 | - | 2 | 178 | ||
AK181112.1 | - | 3 | 220 | ||
BC029106.1 | - | 11 | 1346 | 3711922 |