Gene Transcripts
- Gene Symbol
- Ddx56
- Entrez Gene ID
- 52513
- Gene Name
- DEAD (Asp-Glu-Ala-Asp) box polypeptide 56
- Gene Aliases
- 2600001H07Rik, D11Ertd619e, Noh61
- Chromosome Location
- Chr. 11: 6257545 - 6267729 on Build GRCm38
- UniGene ID
- Mm.29810
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026538.3 |
NP_080814.1
|
11, 12 |
1395 |
|
GenBank mRNA |
AK011136.1 |
- |
11, 12 |
1405 |
|
AK076047.1 |
- |
11, 12 |
1413 |
|
AK076975.1 |
- |
11, 12 |
1395 |
|
AK077514.1 |
- |
11, 12 |
1408 |
|
AK077953.1 |
- |
6, 7 |
642 |
|
AK145628.1 |
- |
11, 12 |
1436 |
|
AK159793.1 |
- |
11, 12 |
1399 |
|
AK169654.1 |
- |
11, 12 |
1410 |
|
AK185833.1 |
- |
3, 4 |
214 |
|
BC018291.1 |
- |
11, 12 |
1389 |
4190167 |
View Full Product Details