Gene Transcripts

Gene Symbol
Ccdc43
Entrez Gene ID
52715
Gene Name
coiled-coil domain containing 43
Gene Aliases
2610001E01Rik, D11Ertd707e
Chromosome Location
Chr. 11: 102684684 - 102697943 on Build GRCm38
UniGene ID
Mm.277638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025918.3 NP_080194.1 4, 5 501
GenBank mRNA AK009319.1 - 4, 5 497
AK011254.1 - 4, 5 502
AK028834.1 - 4, 5 512
AK166820.1 - 4, 5 497
BC061076.1 - 4, 5 510 6594359

View Full Product Details