Gene Transcripts

Gene Symbol
Smarcal1
Entrez Gene ID
54380
Gene Name
SWI/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1
Gene Aliases
6030401P21Rik, AI851716, Mharp
Chromosome Location
Chr. 1: 72536687 - 72636790 on Build GRCm38
UniGene ID
Mm.274232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018817.2 NP_061287.2 5 1260
GenBank mRNA AF088884.1 - 5 1240
AK035854.1 - 5 1237
AK042332.1 - 5 1239
AK077878.1 - 5 1238
AK083488.1 - 5 1238
AK146767.1 - 5 1254
AK154020.1 - 5 1243
AK161570.1 - 5 1180
AK169465.1 - 5 1251
BC029078.1 - 5 1195 3992325

View Full Product Details