Gene Transcripts
- Gene Symbol
- Dll4
- Entrez Gene ID
- 54485
- Gene Name
- delta-like 4 (Drosophila)
- Gene Aliases
- Delta4
- Chromosome Location
- Chr. 2: 119325784 - 119335666 on Build GRCm38
- UniGene ID
- Mm.143719
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019454.3 |
NP_062327.2
|
2, 3 |
705 |
|
GenBank mRNA |
AB043893.1 |
- |
2, 3 |
348 |
|
AF253469.1 |
- |
2, 3 |
660 |
|
AF273454.1 |
- |
2, 3 |
726 |
|
AK004739.1 |
- |
2, 3 |
673 |
|
AK031699.1 |
- |
2, 3 |
674 |
|
AK052322.1 |
- |
2, 3 |
706 |
|
AK165517.1 |
- |
2, 3 |
674 |
|
AK171717.1 |
- |
2, 3 |
674 |
|
BC042497.1 |
- |
2, 3 |
660 |
4017786 |
BC049130.1 |
- |
2, 3 |
665 |
6825525 |
View Full Product Details