Gene Transcripts

Gene Symbol
Htra2
Entrez Gene ID
64704
Gene Name
HtrA serine peptidase 2
Gene Aliases
AI481710, Omi, Prss25, mnd2
Chromosome Location
Chr. 6: 83051266 - 83054571 on Build GRCm38
UniGene ID
Mm.21880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019752.3 NP_062726.3 3 1019
GenBank mRNA AF164513.1 - 3 898
AF175324.1 - 3 915
AK011536.1 - 1825
AK146338.1 - 3 910
AK159188.1 - 3 839
BC033276.1 - 3 879
BC049880.1 - 3 1019 5321861

View Full Product Details