Gene Transcripts

Gene Symbol
Scel
Entrez Gene ID
64929
Gene Name
sciellin
Gene Aliases
9230114I02Rik
Chromosome Location
Chr. 14: 103513060 - 103613346 on Build GRCm38
UniGene ID
Mm.244003
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022886.2 NP_075024.2 17 1128
GenBank mRNA AF245700.2 - 17 1128
BC025142.1 - 17 1100
BC111874.1 - 17 1048 40059586
BC112380.1 - 17 1049 40059585

View Full Product Details