Gene Transcripts

Gene Symbol
Zic5
Entrez Gene ID
65100
Gene Name
zinc finger protein of the cerebellum 5
Gene Aliases
1700049L20Rik, Opr
Chromosome Location
Chr. 14: 122459160 - 122465716 on Build GRCm38
UniGene ID
Mm.390761
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022987.3 NP_075363.1 2 1828
GenBank mRNA AB042155.1 - 2 1850
AB114214.1 - 2 1487
AK006747.1 - 2 226

View Full Product Details