Gene Transcripts

Gene Symbol
Necap2
Entrez Gene ID
66147
Gene Name
NECAP endocytosis associated 2
Gene Aliases
1110005F07Rik, AA409105, C78898
Chromosome Location
Chr. 4: 141066512 - 141078357 on Build GRCm38
UniGene ID
Mm.21187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025383.3 NP_079659.1 2 189
GenBank mRNA AK003463.1 - 2 156
AK145874.1 - 2 177
AK155871.1 - 2 177
AK156566.1 - 2 166
AK170987.1 - 2 174
AK172538.1 - 2 159
AK213029.1 - 2 81
AK218572.1 - 2 98
BB851186.1 - 2 189
BC037069.1 - 2 188 5002765

View Full Product Details