Gene Transcripts

Gene Symbol
Baiap2l1
Entrez Gene ID
66898
Gene Name
BAI1-associated protein 2-like 1
Gene Aliases
1300006M19Rik, AI585895, IRTKS
Chromosome Location
Chr. 5: 144264525 - 144358112 on Build GRCm38
UniGene ID
Mm.18814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025833.3 NP_080109.1 7, 8 916
GenBank mRNA AK004918.2 - 7, 8 917
AK008203.1 - 7, 8 817
BC015459.1 - 7, 8 694 3592632

View Full Product Details