Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166400.1 | NP_001159872.1 | 4 | 931 | |
NM_001166401.1 | NP_001159873.1 | 3 | 903 | ||
NM_005364.4 | NP_005355.2 | 3 | 652 | ||
XM_017029520.1 | XP_016885009.1 | 4 | 719 | ||
GenBank mRNA | AK313943.1 | - | 3 | 652 | |
BC002455.2 | - | 3 | 598 | 3345962 | |
BC012744.2 | - | 3 | 591 | 3617812 | |
BC098120.1 | - | 3 | 527 | ||
BT007340.1 | - | 1 | 382 |