Gene Transcripts

Gene Symbol
Fam103a1
Entrez Gene ID
67148
Gene Name
family with sequence similarity 103, member A1
Gene Aliases
2410047I02Rik, 2610204K14Rik, RAM
Chromosome Location
Chr. 7: 81762953 - 81769490 on Build GRCm38
UniGene ID
Mm.241472
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025997.2 NP_080273.1 3 313
GenBank mRNA AK010694.1 - 3 269
AK011881.1 - 3 313
AK018614.1 - 3 312
AK079819.1 - 3 292
AK081301.1 - 3 305
AK085230.1 - 3 311
AK150938.1 - 3 280
AK160347.1 - 3 312
AK169392.1 - 3 342
BC058367.1 - 3 236 6830048
BC091760.1 - 3 273 5042613
BC096399.1 - 3 271 876882

Gene Symbol
Gm10767
Entrez Gene ID
100038538
Gene Name
predicted gene 10767
Gene Aliases
ENSMUSG00000074826
Chromosome Location
Chr. 13: 66905415 - 66909166 on Build GRCm38
UniGene ID
Mm.405005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177750.1 NP_001171221.1 3 612
GenBank mRNA AK139034.1 - 3 613
AK163421.1 - 959

View Full Product Details