Gene Transcripts

Gene Symbol
Aldh3b1
Entrez Gene ID
67689
Gene Name
aldehyde dehydrogenase 3 family, member B1
Gene Aliases
1700001N19Rik, ALDH4, ALDH7
Chromosome Location
Chr. 19: 3913491 - 3930817 on Build GRCm38
UniGene ID
Mm.109341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026316.2 NP_080592.2 7 727
GenBank mRNA AK154500.1 - 7 773
AK154723.1 - 7 820
AK159447.1 - 7 741
AK160106.1 - 7 744
BC023305.1 - 7 748
BC046597.1 - 7 727 6312211

View Full Product Details