Gene Transcripts

Gene Symbol
Zmym4
Entrez Gene ID
67785
Gene Name
zinc finger, MYM-type 4
Gene Aliases
6330503C17Rik, AI480785, AW493829, CDIR, D630001M21, MYM, Zfp262, Znf262, mKIAA0425
Chromosome Location
Chr. 4: 126861819 - 126969370 on Build GRCm38
UniGene ID
Mm.165401
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114399.1 NP_001107871.1 26, 27 4017
GenBank mRNA AK129137.1 - 26, 27 4272
BC038024.1 - 4, 5 466
BC050924.1 - 20, 21 2926

View Full Product Details