Gene Transcripts

Gene Symbol
Cst13
Entrez Gene ID
69294
Gene Name
cystatin 13
Gene Aliases
1700006C19Rik, Cymg1, Mtcg1
Chromosome Location
Chr. 2: 148820099 - 148830410 on Build GRCm38
UniGene ID
Mm.46167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027024.3 NP_081300.1 2 466
GenBank mRNA AK005665.1 - 2 444
AY600990.2 - 2 484
BC048681.1 - 2 466 6774445

View Full Product Details