Gene Transcripts

Gene Symbol
Cyp2c66
Entrez Gene ID
69888
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 66
Gene Aliases
2010301M18Rik
Chromosome Location
Chr. 19: 39113898 - 39186756 on Build GRCm38
UniGene ID
Mm.271190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011707.1 NP_001011707.1 5 699
GenBank mRNA AK008512.2 - 4 525
AY227734.1 - 5 699
BC147819.1 - 5 696 9087536
BC147824.1 - 5 696 9087541

View Full Product Details