Gene Transcripts
- Gene Symbol
- Phf6
- Entrez Gene ID
- 70998
- Gene Name
- PHD finger protein 6
- Gene Aliases
- 2700007B13Rik, 4931428F02Rik, mKIAA1823
- Chromosome Location
- Chr. X: 52912214 - 52956961 on Build GRCm38
- UniGene ID
- Mm.26870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290546.1 |
NP_001277475.1
|
2, 3 |
363 |
|
NM_027642.2 |
NP_081918.1
|
4, 5 |
649 |
|
GenBank mRNA |
AK016480.1 |
- |
4, 5 |
650 |
|
AK028498.1 |
- |
2, 3 |
285 |
|
AK038320.1 |
- |
4, 5 |
569 |
|
AK122560.1 |
- |
4, 5 |
579 |
|
AK141804.1 |
- |
4, 5 |
592 |
|
AK163345.1 |
- |
4, 5 |
605 |
|
BC043127.1 |
- |
4, 5 |
597 |
6410361 |
BC055330.1 |
- |
4, 5 |
592 |
6406483 |
BC057374.1 |
- |
4, 5 |
555 |
6826091 |
View Full Product Details