Gene Transcripts

Gene Symbol
Lrrc34
Entrez Gene ID
71827
Gene Name
leucine rich repeat containing 34
Gene Aliases
1700007J06Rik, SPATA34
Chromosome Location
Chr. 3: 30624267 - 30647818 on Build GRCm38
UniGene ID
Mm.45373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027941.1 NP_082217.1 3 319
GenBank mRNA AK005720.1 - 3 371
BC100519.1 - 3 319 30919449
BC116419.1 - 2 152
BC116420.1 - 2 159 40103399
JF934738.1 - 3 270

View Full Product Details