Gene Transcripts

Gene Symbol
Mms19
Entrez Gene ID
72199
Gene Name
MMS19 (MET18 S. cerevisiae)
Gene Aliases
2410001K24Rik, 2610042O15Rik, AI316855, C79368, C86341, Mms19l
Chromosome Location
Chr. 19: 41943707 - 41981159 on Build GRCm38
UniGene ID
Mm.218940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028152.3 NP_082428.1 9, 10 793
GenBank mRNA AF319949.1 - 9, 10 787
AK010326.1 - 9, 10 797
AK011754.1 - 9, 10 787
AK028317.1 - 9, 10 793
AK030619.1 - 9, 10 794
AK036983.1 - 9, 10 787
AK050265.1 - 5, 6 1108
AK078688.1 - 9, 10 786
AK087633.1 - 9, 10 750
AK213193.1 - 3, 4 185
BC018614.1 - 9, 10 771
BC050817.1 - 9, 10 773 6332214

View Full Product Details