Gene Transcripts

Gene Symbol
Ccdc74a
Entrez Gene ID
72315
Gene Name
coiled-coil domain containing 74A
Gene Aliases
2310015A05Rik
Chromosome Location
Chr. 16: 17645903 - 17650739 on Build GRCm38
UniGene ID
Mm.297777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166164.1 NP_001159636.1 1, 2 321
GenBank mRNA AK009347.1 - 1, 2 174

View Full Product Details