Gene Transcripts

Gene Symbol
Izumo1
Entrez Gene ID
73456
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
1700058F15Rik, Izumo
Chromosome Location
Chr. 7: 45621811 - 45627242 on Build GRCm38
UniGene ID
Mm.380445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018013.1 NP_001018013.1 7 831
GenBank mRNA AB195681.1 - 7 831
AK006830.1 - 7 831
BC139270.1 - 7 715

View Full Product Details