Gene Transcripts

Gene Symbol
Ms4a13
Entrez Gene ID
73466
Gene Name
membrane-spanning 4-domains, subfamily A, member 13
Gene Aliases
1700060E18Rik, Tetm4.2
Chromosome Location
Chr. 19: 11169418 - 11196755 on Build GRCm38
UniGene ID
Mm.272723
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198224.3 NP_937867.2 5 560
GenBank mRNA AF544402.1 - 5 565
BC089483.1 - 5 560 6772390

View Full Product Details