Gene Transcripts

Gene Symbol
Spata19
Entrez Gene ID
75469
Gene Name
spermatogenesis associated 19
Gene Aliases
1700001L23Rik, AV047838, spergen-1
Chromosome Location
Chr. 9: 27396821 - 27401711 on Build GRCm38
UniGene ID
Mm.45824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB120716.1 - 4 300
AK005610.1 - 4 300
BC049742.1 - 3 244 6771761
JX089334.1 - 4 600

View Full Product Details