Gene Transcripts

Gene Symbol
Vmn2r61
Entrez Gene ID
637873
Gene Name
vomeronasal 2, receptor 61
Gene Aliases
Casr-rs2, EG637873, Gprc2a-rs2
Chromosome Location
Chr. 7: 42260053 - 42300755 on Build GRCm38
UniGene ID
Mm.469899
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105058.1 NP_001098528.1 3 673

Gene Symbol
Vmn2r-ps54
Entrez Gene ID
75835
Gene Name
vomeronasal 2, receptor, pseudogene 54
Gene Aliases
1700108E19Rik, 4930562N12Rik, Casr-rs5, ENSMUSG00000078527, Gm10948, Gprc2a-rs5
Chromosome Location
Chr. 7: 41668014 - 41727193 on Build GRCm38
UniGene ID
Mm.159597
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK007132.1 - 4 322
AK016194.1 - 3 415
AK030272.1 - 3 382

View Full Product Details