Gene Transcripts

Gene Symbol
NFATC1
Entrez Gene ID
4772
Gene Name
nuclear factor of activated T-cells 1
Gene Aliases
NF-ATC, NF-ATc1.2, NFAT2, NFATc
Chromosome Location
Chr. 18: 79395772 - 79529323 on Build GRCh38
UniGene ID
Hs.534074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278669.1 NP_001265598.1 5 2061
NM_001278670.1 NP_001265599.1 5 2061
NM_001278672.1 NP_001265601.1 5 1687
NM_001278673.1 NP_001265602.1 4 588
NM_001278675.1 NP_001265604.1 5 1687
NM_006162.4 NP_006153.2 5 2061
NM_172387.2 NP_765975.1 5 1687
NM_172388.2 NP_765976.1 4 962
NM_172389.2 NP_765977.1 5 1687
NM_172390.2 NP_765978.1 5 2061
XM_017025783.1 XP_016881272.1 5 1903
GenBank mRNA AK292641.1 - 5 1903
AK293760.1 - 4 533
BC104753.1 - 5 1702 8143756
BC112243.1 - 5 1702 8327711
BC143457.1 - 5 1702
EU887559.1 - 5 1608
EU887560.1 - 5 1569
EU887561.1 - 5 1608
EU887562.1 - 5 1569
EU887563.1 - 5 1608
EU887564.1 - 5 1569
EU887565.1 - 5 1608
EU887566.1 - 5 1569
U08015.1 - 5 1847
U59736.1 - 5 1644
U80917.1 - 5 1977
U80918.1 - 5 1631
U80919.1 - 4 790

View Full Product Details