Gene Transcripts

Gene Symbol
Arid2
Entrez Gene ID
77044
Gene Name
AT rich interactive domain 2 (ARID, RFX-like)
Gene Aliases
1700124K17Rik, 4432409D24Rik, zipzap/p200
Chromosome Location
Chr. 15: 96287522 - 96405463 on Build GRCm38
UniGene ID
Mm.17166
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175251.4 NP_780460.3 15 3112
GenBank mRNA AK014481.1 - 2 1077
AK173201.1 - 1 939
BC027180.1 - 11 4021271

View Full Product Details