Gene Transcripts

Gene Symbol
TONSL
Entrez Gene ID
4796
Gene Name
tonsoku-like, DNA repair protein
Gene Aliases
IKBR, NFKBIL2
Chromosome Location
Chr. 8: 144428780 - 144444488 on Build GRCh38
UniGene ID
Hs.675285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013432.4 NP_038460.4 11 1394
XM_011517048.2 XP_011515350.1 4 493
XM_011517049.2 XP_011515351.1 4 454
XM_011517050.2 XP_011515352.1 11 1453
GenBank mRNA AB209126.1 - 3 2354
BC008782.2 - 10 1366 3628374

View Full Product Details