Gene Transcripts

Gene Symbol
Fbxo43
Entrez Gene ID
78803
Gene Name
F-box protein 43
Gene Aliases
4930533G20Rik, Emi2
Chromosome Location
Chr. 15: 36150060 - 36173980 on Build GRCm38
UniGene ID
Mm.26393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081253.1 NP_001074722.1 2 1162
GenBank mRNA AK019714.1 - 2 1098
AK030012.1 - 2 986
BC098484.1 - 2 1162 30053208

View Full Product Details