Gene Transcripts

Gene Symbol
Herpud2
Entrez Gene ID
80517
Gene Name
HERPUD family member 2
Gene Aliases
5031400M07Rik, AB041580
Chromosome Location
Chr. 9: 25108124 - 25151781 on Build GRCm38
UniGene ID
Mm.142843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020586.2 NP_065611.1 3 1120
GenBank mRNA AB041580.1 - 3 915
AK019862.1 - 3 1065
AK135935.1 - 3 1121
BC029691.1 - 3 338 5375603
BC043693.1 - 3 1159 5150485

View Full Product Details